Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, Decahydrate, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7727-73-3
H20Na2O14S
322.19
RSIJVJUOQBWMIM-UHFFFAOYSA-L
disodium decahydrate sulfate
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O
7727-73-3
H20Na2O14S
322.19
RSIJVJUOQBWMIM-UHFFFAOYSA-L
disodium decahydrate sulfate
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O

S1458, 7727-73-3, Na2SO4•10H2O

Spectrum™ Sodium Sulfate, Decahydrate, Reagent, ACS is the sodium salt of sulfuric acid. It is a white, crystalline solid, and is used mainly in the manufacture of detergents. Sodium sulfate has been used in the laboratory for protein precipitation. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Grade ACS
CAS Max % 1
Percent Purity 99%
Calcium (Ca) 5e-005
Heavy Metals (as Pb) 3 ppm
Insoluble Matter 0.0001
Iron (Fe) 5 ppm
Magnesium (Mg) 3e-005
Packaging Poly Pail
Potassium (K) 5e-005
pH 5.2 to 9.2
Quantity 12 kg
Show More Show Less