Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfathiazole, Sesquihydrate, 99%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle

S1462, C9H8N3NaO2S2•1.5H2O

Spectrum™ Sodium Sulfathiazole, Sesquihydrate , an organosulfur compound, can be used as a short-acting sulfa drug and as an oral and topical antimicrobial, Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Grade Reagent
CAS Max % 1
Percent Purity 99%
Loss on Drying 0.09
Packaging Poly Bottle
Quantity 500 g