Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide, 9-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
1313-84-4
Na2S·9H2O
1313-84-4
Na2S·9H2O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 240.18
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.43kg
Quantity 500g
Chemical Name or Material Sodium Sulfide, 9-Hydrate