Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide, Fused Flakes, Approx. 60%, Spectrum™

Click to view available options
Quantity:
12 kg
2.5 kg
400 lbs.
500 g
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
27610-45-3
H3Na2OS
97.06
BNJIAWKLYDUTBN-UHFFFAOYSA-M
disodium hydrate sulfanide
O.[Na+].[Na+].[SH-]
27610-45-3
H3Na2OS
97.06
BNJIAWKLYDUTBN-UHFFFAOYSA-M
disodium hydrate sulfanide
O.[Na+].[Na+].[SH-]

S1470, 27610-45-3, Na2S•xH2O

Spectrum™ Sodium Sulfide, Fused Flakes , also known as disodium sulfide, is used as an oxygen scavenger agent in water treatment as well as a metals precipitant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Grade Reagent
CAS Max % 1
Percent Purity Approx. 60%
Packaging Poly Pail
Quantity 12 kg