Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfide hydrate, 60-64%, Acros Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Fibre drum
Plastic bottle
27610-45-3
Na2S·xH2O
78.04
MFCD00149183
BNJIAWKLYDUTBN-UHFFFAOYSA-M
45051681
disodium;sulfanide;hydrate
O.[Na+].[Na+].[SH-]
27610-45-3
Na2S·xH2O
78.04
MFCD00149183
BNJIAWKLYDUTBN-UHFFFAOYSA-M
45051681
disodium;sulfanide;hydrate
O.[Na+].[Na+].[SH-]
Assay Percent Range 60-64%
Linear Formula Na2S·xH2O
Arsenic (As) 1ppm max.
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol,insoluble in ether
Formula Weight 78.04
Physical Form Flakes and Chunks
Percent Purity 60-64%
Grade Extra Pure
Heavy Metals (as Pb) 2ppm max.
Iron (Fe) 10ppm max.
Packaging Plastic bottle
Polysulfides < 0.4%
Sodium Carbonate 0.7% max. (Na2CO3)
Sodium Hydrosulfide < 2%
Sodium Thiosulfate < 0.7%
Sodium Sulfite < 0.6%
Solubility Solubility in water: soluble. Other solubilities: slightly soluble in alcohol, insoluble in ether
Trace Analysis 2ppm max.
Color Yellow
Quantity 1kg
Chemical Name or Material Sodium sulfide hydrate
Show More Show Less

GHS P Statement
May be corrosive to metals.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger