Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide, Nonahydrate, Crystal, Reagent, ACS, 98%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
Packaging:
Amber Glass Bottle
1313-84-4
H18Na2O9S
240.17
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
disodium nonahydrate sulfanediide
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
1313-84-4
H18Na2O9S
240.17
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
disodium nonahydrate sulfanediide
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]

S1465, 1313-84-4, Na2S•9H2O

Spectrum™ Sodium Sulfide, Nonahydrate, Crystal, Reagent, ACS is a colorless chemical compound that is soluble in water. It often appears yellow in color due to the presence of polysulfides, although it is colorless as a solution. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Grade ACS
CAS Max % 1
Percent Purity 98%
Iron (Fe) To pass test
Packaging Amber Glass Bottle
Quantity 125 g