Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium sulfide, tech., 90+%, H2O 5% max

Manufacturer:  Alfa Aesar™ 8869709

Quantity 10g

 View more versions of this product

Catalog No. AA8869709


  Check Availability
Add to cart

Specifications

Description

  • For reducing organic nitro compounds, in the manufacture of sulfur dyes, in ore flotation, and in the preparation of metal sulfide finishes
Description & Specifications

Chemical Identifiers

12034-39-8
174.24
UN1385
Technical Grade
Na2S4
MFCD00014249
Sodium sulfide

Specifications

12034-39-8
Na2S4
MFCD00014249
Soluble in water and alcohol
Like rotten eggs
Pale brown
10g
≥90%
Powder
174.24
UN1385
H2O 5% max
Moisture Sensitive & Hygroscopic
275°C
Sodium sulfide
Technical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H252-H311-H314-H318-H302
Flammable solid.
Self-heating in large quantities
may catch fire.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280-P273-P305+P351+P338-P309-P310-P420
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store away from other materials.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: SODIUM SULFIDE, ANHYDROUS

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit