Alfa Aesar™ Sodium sulfide, tech., 90+%, H2O 5% max

Manufacturer: Alfa Aesar™ 8869718

Quantity 50g

 View more versions of this product

Catalog No. AA8869718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12034-39-8
    Molecular Formula Na2S4
    Formula Weight 174.24
    MDL Number MFCD00014249
    UN Number UN1385
    Chemical Name or Material Sodium sulfide
    Grade Technical Grade

    Specifications

    Formula Weight 174.24
    Health Hazard 3 GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P420
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store away from other materials.
    Health Hazard 2 GHS H Statement
    H228-H252-H311-H314-H318-H302
    Flammable solid.
    Self-heating in large quantities
    may catch fire.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Physical Form Powder
    Health Hazard 1 Danger
    Concentration H2O 5% max
    Molecular Formula Na2S4
    MDL Number MFCD00014249
    CAS 12034-39-8
    UN Number UN1385
    Color Pale brown
    Solubility Information Soluble in water and alcohol
    DOT Information Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: SODIUM SULFIDE, ANHYDROUS
    TSCA TSCA
    Sensitivity Moisture Sensitive & Hygroscopic
    Quantity 50g
    Odor Like rotten eggs
    Melting Point 275°C
    Chemical Name or Material Sodium sulfide
    Assay ≥90%
    Grade Technical Grade

    Description

    • For reducing organic nitro compounds, in the manufacture of sulfur dyes, in ore flotation, and in the preparation of metal sulfide finishes

    GHS H Statement
    H228-H252-H311-H314-H318-H302
    Flammable solid.
    Self-heating in large quantities
    may catch fire.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P420
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store away from other materials.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit