Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfite, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

7757-83-7
Na2O3S
126.037
GEHJYWRUCIMESM-UHFFFAOYSA-L
24437
CHEBI:86477

High quality chemicals for laboratory and specialized industrial use

SMILES [O-]S(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;sulfite
Formula Weight 126.04
Packaging Poly Bottle
Specific Gravity 1L = 2.60kg
Quantity 2.5kg
Chemical Name or Material Sodium Sulfite, Anhydrous
Grade Baker Analyzed™ ACS Reagent