Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfite, NF, FCC, EP, BP, 95-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7757-83-7
H14Na2O10S
252.14
WMJRPJZQQSSDBU-UHFFFAOYSA-L
disodium heptahydrate sulfite
O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O
7757-83-7
H14Na2O10S
252.14
WMJRPJZQQSSDBU-UHFFFAOYSA-L
disodium heptahydrate sulfite
O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])=O

S1113, 7757-83-7, Na2SO3

Spectrum™ Sodium Sulfite, NF, FCC, EP, BP is a widely used preservative. All Spectrum Chemical NF, FCC, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade Multi Compendia
CAS Max % 1
Percent Purity 95 to 100.5%
Iron (Fe) 10 ppm
Lead (Pb) 2 mg/kg
Packaging Poly Bottle
Selenium (Se) 10 ppm
Zinc (Zn) 25 ppm
Quantity 2.5 kg