Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tartrate Dihydrate, ACS Reagent, Reagents

Catalog No. 01337665
Change view
Click to view available options
Quantity:
125 g
500 g
2 kg
12 kg
Catalog No. Quantity
01-337-665 2 kg
01-337-667 125 g
01-337-666 12 kg
01-337-664 500 g
4 options

Catalog No. 01-337-665

Supplier: Reagents Holdings Llc C13270002A5

Only null left
Add to Cart
Add to Cart

Sodium Tartrate Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Melting Point 150°C
Color Colorless/White
pH 7 to 9
Physical Form Solid
Quantity 2 kg
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.82
Chemical Name or Material Sodium Tartrate Dihydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.