Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tartrate, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

6106-24-7
C4H10Na2O8
232.096
YAPIZISBZUXUIH-DGFHWNFOSA-N
131855972
(2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
C(C(C(=O)O)O)(C(=O)O)O.O.O.[Na].[Na]

High quality chemicals for laboratory and specialized industrial use

    Suitable for Standardization of Karl Fischer Reagents
Quantity 2.5kg
Specific Gravity 1L = 1.82kg
Packaging Poly Bottle
Formula Weight 230.08
Physical Form Crystals
Chemical Name or Material Sodium Tartrate, Dihydrate
Grade Baker Analyzed™ ACS Reagent