Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tartrate TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Poly Bottle
6106-24-7,7742-18-5
C4H10Na2O8
232.10
YAPIZISBZUXUIH-UHFFFAOYNA-N
2,3-dihydroxybutanedioic acid dihydrate disodium
O.O.[Na].[Na].OC(C(O)C(O)=O)C(O)=O
6106-24-7,7742-18-5
C4H10Na2O8
232.10
YAPIZISBZUXUIH-UHFFFAOYNA-N
2,3-dihydroxybutanedioic acid dihydrate disodium
O.O.[Na].[Na].OC(C(O)C(O)=O)C(O)=O

S-214, 6106-24-7

Spectrum™ Sodium Tartrate TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

CAS Max % 0.001,0.999
Grade Chemical Solution
Packaging Poly Bottle
Quantity 100 mL