Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tellurate(VI) hydrate, Te 45% min, Thermo Scientific Chemicals

Catalog Number p-7080338
Click to view available options
Quantity:
10 g
50 g
10101-83-4
Na2O4Te
237.58
MFCD00014242
XERQTZLDFHNZIC-UHFFFAOYSA-L
sodium tellurate, unii-7kar02f0r0, natriumtellurat, 2na.o4te, disodium tellurate, telluric acid h2teo4 , sodium salt 1:2, telluric acid h2teo4 ,disodium salt, dihydrate 8ci,9ci
13652393
[Na+].[Na+].[O-][Te]([O-])(=O)=O
10101-83-4
Na2O4Te
237.58
MFCD00014242
XERQTZLDFHNZIC-UHFFFAOYSA-L
sodium tellurate, unii-7kar02f0r0, natriumtellurat, 2na.o4te, disodium tellurate, telluric acid h2teo4 , sodium salt 1:2, telluric acid h2teo4 ,disodium salt, dihydrate 8ci,9ci
13652393
[Na+].[Na+].[O-][Te]([O-])(=O)=O

Sodium tellurite is an intermediate in the extraction of the element, tellurium; it is a product obtained from anode slimes and is a precursor to tellurium. Solutions of sodium tellurite are used for black or blue-black coatings on iron, steel, aluminum, and copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Physical Form Crystalline
Quantity 10 g
Merck Index 14,8686
Solubility Information Soluble in water.
IUPAC Name disodium tellurate
Formula Weight 237.58 (Anhydrous)
Concentration or Composition (by Analyte or Components) Te 45% min
Chemical Name or Material Sodium tellurate (VI) hydrate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 233-259-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit