Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetraborate, Anhydrous, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 12305A1

Quantity 1kg

 View more versions of this product

Catalog No. AA12305A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1330-43-4
99.5% (metals basis)
201.22
Borax, anhydrous
Granular
B4Na2O7
MFCD00081185
Sodium tetraborate, anhydrous

Specifications

1330-43-4
99.5% (metals basis)
White powder
MFCD00081185
Odorless
Green
Sodium tetraborate, anhydrous
Granular
B4Na2O7
201.22
Borax, anhydrous
0 hPa
1kg

Safety and Handling

SDS & Safety Documents

H360: May damage fertility or the unborn child.

EINECSNumber : 215-540-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit