Sodium tetraborate, anhydrous, 99.95% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 8786222

Quantity 100g

 View more versions of this product

Catalog No. AA8786222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1330-43-4
    Physical Form Powder
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula B4Na2O7
    Formula Weight 201.22
    MDL Number MFCD00081185
    Synonym Borax
    Chemical Name or Material Sodium tetraborate

    Specifications

    Physical Form Powder
    Formula Weight 201.22
    Quantity 100g
    Color White
    EINECS Number 215-540-4
    Anhydrous or Hydrate Anhydrous
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula B4Na2O7
    MDL Number MFCD00081185
    CAS 1330-43-4
    Synonym Borax
    Health Hazard 1 H360: May damage fertility or the unborn child.
    Odor Odorless
    Health Hazard 2 Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    Chemical Name or Material Sodium tetraborate

    Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H360: May damage fertility or the unborn child.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit