Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetraborate, anhydrous, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8786214

Quantity 25g

 View more versions of this product

Catalog No. AA8786214


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1330-43-4
99.95% (metals basis)
201.22
Borax
Powder
B4Na2O7
MFCD00081185
Sodium tetraborate

Specifications

1330-43-4
99.95% (metals basis)
201.22
Borax
Odorless
White
Sodium tetraborate
Powder
B4Na2O7
MFCD00081185
2.637g/cm3 at 20°C
Anhydrous
25g

Safety and Handling

SDS & Safety Documents

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H360: May damage fertility or the unborn child.

EINECSNumber : 215-540-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit