Learn More
Sodium tetraborate, anhydrous, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Sodium tetraborate is used as a co-catalyst for (2,2,6,6-tetramethylpiperidin-1-yl)oxyl and sodium hypochlorite oxidation of alcohols in greener and non-chlorinated solvents. It acts as a reagent for the decomposition of oxides and silicates by fusion. It is used to prepare buffers, which is used in gel electrophoresis of DNA. It acts as a source of borate, micronutrient fertilizer, preservative in taxidermy, additive in ceramic slips and glazes. It is also used as a precursor for sodium perborate monohydrate and boric acid. It finds application as fluoride detoxification, neutron absorber in nuclear reactors and to make indelible ink for dip pens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1575°C (decomposition) |
Melting Point | 741°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Linear Formula | Na2B4O7 |
Merck Index | 14,8590 |
Solubility Information | Soluble in water and glycerol. Slightly soluble in ethanol. Insoluble in acids. |
Formula Weight | 201.22 |
Percent Purity | 99.998% |
Show More |
Safety and Handling
Hazard Category | H319-H360FD |
Hazard Statement | GHS H Statement H360 May damage fertility or the unborn child. |
Precautionary Statement | P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c |
EINECSNumber | 215-540-4 |
RTECSNumber | ED4588000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.