Sodium tetrabromoaurate(III) hydrate, 99.99% (Metals basis), Alfa Aesar™, Premion™

Manufacturer: Alfa Aesar 4042806

Quantity 5g

 View more versions of this product

Catalog No. AA4042806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10378-49-1
    Physical Form Crystalline Powder
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula NaAuBr4·xH2O (x?2)
    Formula Weight 539.59
    MDL Number MFCD00274624
    Chemical Name or Material Sodium tetrabromoaurate(III) hydrate

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 539.59
    Quantity 5g
    Odor Odorless
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula NaAuBr4·xH2O (x?2)
    Appearance Crystalline powder
    MDL Number MFCD00274624
    CAS 10378-49-1
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing.
    Chemical Name or Material Sodium tetrabromoaurate(III) hydrate

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing.

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit