Sodium tetrachloroaluminate, Alfa Aesar™

Manufacturer: Alfa Aesar 2237506

Quantity 5g

 View more versions of this product

Catalog No. AA2237506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-16-9
    Physical Form Powder
    Molecular Formula AlCl4Na
    Formula Weight 191.78
    MDL Number MFCD00050361
    Synonym Aluminum sodium chloride
    Chemical Name or Material Sodium tetrachloroaluminate

    Specifications

    Physical Form Powder
    Formula Weight 191.78
    Quantity 5g
    Color Pale yellow
    DOT Information Hazard Class: 8, Packing Group: III
    EINECS Number 232-050-6
    Molecular Formula AlCl4Na
    Appearance Pale yellow powder
    MDL Number MFCD00050361
    CAS 7784-16-9
    Synonym Aluminum sodium chloride
    Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Sodium tetrachloroaluminate

    H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit