Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium tetrachloropalladate(II), 99.9% (metals basis), Pd 35.4% min

Manufacturer:  Alfa Aesar™ 4200009

Quantity 10g

 View more versions of this product

Catalog No. AA4200009

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

13820-53-6
294.19
MFCD00003487
Sodium tetrachloropalladate(II)

Specifications

13820-53-6
294.19
Pd 35.4% min.
10g
99.9%, (metals basis)
Crystalline
MFCD00003487
Red-brown
Sodium tetrachloropalladate(II)

Safety and Handling

GHS H Statement
H318-H290-H302
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.

GHS P Statement
P280f-P305+P351+P338
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

Danger

RTECSNumber : RT3480000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit