Alfa Aesar™ Sodium tetrachloropalladate(II), 99.9% (metals basis), Pd 35.4% min

Manufacturer: Alfa Aesar™ 42000-04

Quantity 2g

 View more versions of this product

Catalog No. AA4200004

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13820-53-6
    Formula Weight 294.19
    MDL Number MFCD00003487
    Chemical Name or Material Sodium tetrachloropalladate(II)

    Specifications

    Formula Weight 294.19
    Quantity 2g
    Health Hazard 3 GHS P Statement
    P280f-P305+P351+P338
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    TSCA TSCA
    Physical Form Crystalline
    MDL Number MFCD00003487
    CAS 13820-53-6
    Concentration Pd 35.4% min.
    Color Red-brown
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H290-H302
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.
    RTECS Number RT3480000
    Chemical Name or Material Sodium tetrachloropalladate(II)
    Assay 99.9%, (metals basis)

    GHS H Statement
    H318-H290-H302
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.

    GHS P Statement
    P280f-P305+P351+P338
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit