Alfa Aesar™ Sodium tetrachloropalladate(II) hydrate, 99.95% (metals basis), Pd 30%

Manufacturer: Alfa Aesar™ 1188609

Quantity 10g

 View more versions of this product

Catalog No. AA1188609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13820-53-6
    Linear Formula Na2PdCl4·xH2O (x≈3)
    Formula Weight 294.20 (Anhydrous)
    MDL Number MFCD00003487
    Synonym Sodium chloropalladate(II); Palladium sodium chloride
    Chemical Name or Material Sodium tetrachloropalladate(II) hydrate

    Specifications

    Formula Weight 294.20 (Anhydrous)
    Health Hazard 2 GHS H Statement
    H318-H290-H302
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280f-P305+P351+P338
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    TSCA TSCA
    Synonym Sodium chloropalladate(II); Palladium sodium chloride
    Color Red-brown
    Concentration Pd 30%
    Quantity 10g
    RTECS Number RT3480000
    Physical Form Crystalline
    Linear Formula Na2PdCl4·xH2O (x≈3)
    MDL Number MFCD00003487
    CAS 13820-53-6
    Sensitivity Hygroscopic
    Health Hazard 1 Danger
    Chemical Name or Material Sodium tetrachloropalladate(II) hydrate
    Assay 99.95%, (metals basis)

    GHS H Statement
    H318-H290-H302
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.

    GHS P Statement
    P280f-P305+P351+P338
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit