Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetrachloropalladate(II) hydrate, 99.95% (metals basis), Pd 30%, Thermo Scientific Chemicals

Catalog Number p-7048895
Click to view available options
Quantity:
2 g
10 g
13820-53-6
Cl4Na2Pd
294.20
MFCD00003487
ABKQFSYGIHQQLS-UHFFFAOYSA-J
sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
11000870
disodium;tetrachloropalladium(2-)
[Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl
13820-53-6
Cl4Na2Pd
294.20
MFCD00003487
ABKQFSYGIHQQLS-UHFFFAOYSA-J
sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
11000870
disodium;tetrachloropalladium(2-)
[Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl

Sodium tetrachloropalladate is used as a test salt for Pd allergy patch testing. It is used in chemical synthesis as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red to Brown
Linear Formula Na2PdCl4·xH2O (x≈3)
Quantity 2 g
Sensitivity Hygroscopic
Solubility Information Soluble in water and acetic acid.
Formula Weight 294.20 (Anhydrous)
Percent Purity 99.95%
Concentration or Composition (by Analyte or Components) Pd 30%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Sodium tetrachloropalladate(II) hydrate
Show More Show Less
Hazard Category H290-H302-H319
Hazard Statement GHS H Statement
H318-H290-H302
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
Precautionary Statement P234-P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 237-502-6
RTECSNumber RT3480000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit