Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tetraethylborate, 97%, pure, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
15523-24-7
C8H20BNa
150.04
MFCD00061547
SZSBMTRYJRHYNI-UHFFFAOYSA-N
sodium tetraethylborate,sodium tetraethylboranuide,borate 1-,tetraethyl-, sodium 1:1,sodium tetraethylborate 1-,et4bna,sodiotetraethylboron v,borate 1-, tetraethyl-, sodium,sodium tetraethylborate 1g
23681030
sodium;tetraethylboranuide
[B-](CC)(CC)(CC)CC.[Na+]
15523-24-7
C8H20BNa
150.04
MFCD00061547
SZSBMTRYJRHYNI-UHFFFAOYSA-N
sodium tetraethylborate,sodium tetraethylboranuide,borate 1-,tetraethyl-, sodium 1:1,sodium tetraethylborate 1-,et4bna,sodiotetraethylboron v,borate 1-, tetraethyl-, sodium,sodium tetraethylborate 1g
23681030
sodium;tetraethylboranuide
[B-](CC)(CC)(CC)CC.[Na+]
Infrared Spectrum Authentic
Color White
Quantity 5g
Packaging Glass bottle
Melting Point 140.0°C to 142.0°C
Assay Percent Range 97%
Solubility Information Solubility in water: reacts
Formula Weight 150.04
Physical Form Crystalline Powder or Crystals
Percent Purity 97%
Grade Pure
Chemical Name or Material Sodium tetraethylborate
Show More Show Less

GHS H Statement
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.

GHS P Statement
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protecti

GHS Signal Word: Danger