Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiocyanate, 0.100 Normal (N/10), Reagents

Catalog No. 01333543
Change view
Click to view available options
Quantity:
1 L
4 L
Catalog No. Quantity
01-333-543 1 L
01-333-542 4 L
2 options

Catalog No. 01-333-543

Supplier: Reagents Holdings Llc CS1998001A

Only null left
Add to Cart
Add to Cart

Sodium Thiocyanate, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 540-72-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sodium rhodanate, Sodium rhodanide, Sodium sulfocyanate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0019
Chemical Name or Material Sodium Thiocyanate, 0.100 Normal (N/10)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.