Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium thiocyanate, 98%, extra pure, ACROS Organics™

Click to view available options
Quantity:
250g
500g
2.5kg
Packaging:
Plastic bottle
540-72-7
CNNaS
81.07
MFCD00011123
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
516871
CHEBI:30952
sodium;thiocyanate
[Na+].[S-]C#N
540-72-7
CNNaS
81.07
MFCD00011123
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
516871
CHEBI:30952
sodium;thiocyanate
[Na+].[S-]C#N
Assay Percent Range 97.5% min. (on dried substance) (Argentometry)
Linear Formula NaSCN
Fieser 01,1105
Merck Index 15, 9480
Solubility Information Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol,freely soluble in acetone
Formula Weight 81.07
Physical Form Crystalline Powder or Crystals and Chunks
Percent Purity 98%
Grade Extra Pure
Iron (Fe) 2ppm max.
Loss on Drying 2.5% max.
Packaging Plastic bottle
Solubility Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol, freely soluble in acetone
Color White
Melting Point 287.0°C
pH 5.5 to 7.5 (5% soln. at 20°C)
Quantity 250g
Chemical Name or Material Sodium thiocyanate, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
Contact with acids liberates very toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-754-4

RTECSNumber : XL2275000

TSCA : TSCA