Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.01N, LaMotte™ Available on GSA/VA Contract for Federal Government customers only.

Manufacturer:  LaMotte CompanySupplier Diversity Partner 3819H

Quantity 3800mL

 View more versions of this product

Catalog No. 22-278-631


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10102-17-7,497-19-8
Sodium Thiosulfate

Specifications

10102-17-7,497-19-8
Water
Pale yellow
Sodium Thiosulfate
Sodium thiosulfate,Sodium carbonate
Liquid
3800mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit