Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 0.100 Normal Standardized, NIST Traceable, ISO 17025, Reagents

Catalog No. 01336237
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
20 L
Packaging:
Cubitainer
Glass Bottle
Hedpak
Poly Bottle
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-237 20 L Hedpak
01-336-241 10 L Cubitainer
01-336-240 1 L Poly Bottle
01-336-239 1 L Glass Bottle
01-336-238 2.5 L Glass Bottle
01-336-236 20 L Cubitainer
01-336-235 4 L Poly Bottle
01-336-234 4 L Glass Bottle
01-336-233 4 L Cubitainer
01-336-232 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 01-336-237 Supplier Reagents Holdings Llc Supplier No. C732810020E3
Only null left
Add to Cart
Add to Cart

Sodium Thiosulfate, 0.100 Normal Standardized, NIST Traceable, ISO 17025, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Sodium Thiosulfate, 0.100 Normal Standardized, NIST Traceable, ISO 17025
Boiling Point 100°C
Color Colorless
CAS 7732-18-5, 10102-17-7, 10102-17-7
Physical Form Liquid
Quantity 20 L
Molecular Formula Na2S2O3/H2O
Synonym Sodium hyposulfite, Hypo, Hyposulfite of soda
Grade Reagent
Packaging Hedpak
Specific Gravity 1.009
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.