Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 1.00 N, Reagents

Catalog No. 01333749
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
Catalog No. Quantity
01-333-749 10 L
01-333-748 1 L
01-333-747 4 L
3 options

Catalog No. 01-333-749

Supplier: Reagents Holdings Llc CS12660010F

Only null left
Add to Cart
Add to Cart

Sodium Thiosulfate, 1.00 N, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 10102-17-7 , 497-19-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sodium hyposulfite, Hypo, Hyposulfite of soda
IUPAC Name water
SMILES O

Specifications

Boiling Point 101°C
Color Colorless
Physical Form Liquid
Quantity 10 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.1036
Chemical Name or Material Sodium Thiosulfate, 1.00 N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.