Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate, 1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle

Chemical Identifiers

7772-98-7
Na2O3S2
158.097
AKHNMLFCWUSKQB-UHFFFAOYSA-L
24477

High quality chemicals for laboratory and specialized industrial use

CAS 7772-98-7
Molecular Formula Na2O3S2
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES [O-]S(=O)(=S)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Molecular Weight (g/mol) 158.097
PubChem CID 24477
Formula Weight 158.11
Packaging Cubetainer
Specific Gravity 1L = 1.12kg
Quantity 4L
Chemical Name or Material Sodium Thiosulfate, 1N Volumetric Solution
Grade BAKER ANALYZED™ Reagent
Show More Show Less