Alfa Aesar™ Sodium thiosulfate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Manufacturer: Alfa Aesar™ 88628LS

Quantity 6 units

 View more versions of this product

Catalog No. AA88628LS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7772-98-7
    Molecular Formula Na2O3S2
    Linear Formula Na2S2O3
    Formula Weight 158.1
    MDL Number MFCD00003499
    Chemical Name or Material Sodium thiosulfate Acculute Standard Volumetric Solution, Final Concentration 0.1N

    Specifications

    Formula Weight 158.1
    TSCA TSCA
    Physical Form Liquid
    Quantity 6 units
    Molecular Formula Na2O3S2
    Linear Formula Na2S2O3
    MDL Number MFCD00003499
    CAS 7772-98-7
    Chemical Name or Material Sodium thiosulfate Acculute Standard Volumetric Solution, Final Concentration 0.1N

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit