Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium thiosulfate pentahydrate, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
25kg
Packaging:
Fibre drum
Plastic bottle
10102-17-7
Na2O3S2·5H2O
248.18
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate,ametox,sodium thiosulfate, pentahydrate,antichlor,tinver,disodium thiosulfate pentahydrate,unii-hx1032v43m,ccris 3952,thiosulfuric acid, disodium salt, pentahydrate,sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
10102-17-7
Na2O3S2·5H2O
248.18
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate,ametox,sodium thiosulfate, pentahydrate,antichlor,tinver,disodium thiosulfate pentahydrate,unii-hx1032v43m,ccris 3952,thiosulfuric acid, disodium salt, pentahydrate,sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
Reduction reagent
Assay Percent Range 99+%
Linear Formula Na2S2O3·5H2O
Merck Index 15, 8821
Solubility Information Solubility in water: 680g/L (20°C). Other solubilities: practically insoluble in alcohol,soluble in turpentine
Formula Weight 248.18
Physical Form Crystals
Percent Purity ≥99%
Grade Extra Pure
Calcium (Ca) 50ppm max.
Chlorine (Cl) 500ppm max.
Density 1.7200g/mL
Iron (Fe) 10ppm max.
Lead (Pb) 5ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 680g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in turpentine
Specific Gravity 1.72
Sulfate and Sulfite 0.2% max.
Sulfur (S) 5ppm max.
Color Colorless to White
Melting Point 48.5°C
pH 6 to 8.4 (10% soln. at 20°C)
Quantity 1kg
Chemical Name or Material Sodium thiosulfate pentahydrate
Show More Show Less