Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium thiosulfate pentahydrate, ACS Reagent, ≥99.5%, Honeywell Fluka™

Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
10102-17-7
H10Na2O8S2
248.172
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
10102-17-7
H10Na2O8S2
248.172
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
CAS Max % 100.0000%
Quantity 500 g
Linear Formula Na2S2O3 · 5H2O
UN Number NONH for all modes of transport
Solubility Information Soluble in water
Formula Weight 248.18
Percent Purity ≥99.5%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.73 g/cm3
Chemical Name or Material Sodium thiosulfate pentahydrate, ACS Reagent, ≥99.5%
Show More Show Less

ShelfLife : 1080 days from date of manufacture

EINECSNumber : 231-867-5

Recommended Storage : Room Temp