Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate Pentahydrate, Purified, Reagents

Catalog No. 01336949
Click to view available options
Quantity:
500 g
3 kg
10 kg

Sodium Thiosulfate Pentahydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym Sodium hyposulfite, Hypo, Hyposulfite of soda
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Boiling Point 100°C
Melting Point 48°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.69
Chemical Name or Material Sodium Thiosulfate Pentahydrate, Purified
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.