Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Thiosulfate Pentahydrate, Purified, Reagents

Catalog No. 01336950
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
Catalog No. Quantity
01-336-950 3 kg
01-336-951 10 kg
01-336-949 500 g
3 options

Catalog No. 01-336-950

Supplier: Reagents Holdings Llc C23280003A5

Only null left
Add to Cart
Add to Cart

Sodium Thiosulfate Pentahydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym Sodium hyposulfite, Hypo, Hyposulfite of soda
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Boiling Point 100°C
Melting Point 48°C
Color Colorless/White
Physical Form Solid
Quantity 3 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.69
Chemical Name or Material Sodium Thiosulfate Pentahydrate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.