Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium thiosulfate pentahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Click to view available options
Quantity:
500g
1kg
5kg
6 x 1kg
10102-17-7
H10Na2O8S2
248.172
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate,ametox,sodium thiosulfate, pentahydrate,antichlor,tinver,disodium thiosulfate pentahydrate,unii-hx1032v43m,ccris 3952,thiosulfuric acid, disodium salt, pentahydrate,sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
10102-17-7
H10Na2O8S2
248.172
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate,ametox,sodium thiosulfate, pentahydrate,antichlor,tinver,disodium thiosulfate pentahydrate,unii-hx1032v43m,ccris 3952,thiosulfuric acid, disodium salt, pentahydrate,sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula Na2S2O3 · 5H2O
UN Number NONH for all modes of transport
Formula Weight 248.18g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Quantity 5kg
Chemical Name or Material Sodium Thiosulfate Pentahydrate, Reag. Ph. Eur. reag. ISO

EINECSNumber : 231-867-5