Learn More
Sodium triacetoxyborohydride, 95%, Thermo Scientific Chemicals
Description
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.
Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
Specifications
Melting Point | ∼110°C (decomposition) |
Quantity | 5 g |
UN Number | UN3396 |
Beilstein | 4047608 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in dimethyl sulfoxide,methanol,benzene,toluene,terahydrofuran,dioxane and methylene chloride. |
Formula Weight | 211.94 |
Percent Purity | 95% |
Chemical Name or Material | Sodium triacetoxyborohydride |
Safety and Handling
Hazard Category | H260-H315-H318 |
Hazard Statement | GHS H Statement H261-H228-H318-H335-H315 In contact with water releases flammable gas. Flammable solid. Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
Precautionary Statement | P223-P231+P232-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P335+P334-P362-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.3; Packaging Group: II |
EINECSNumber | 000-000-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.