Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium triethylborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
17979-81-6, 109-99-9
C6H16BNa
121.99
MFCD00011704
UKYGKELSBAWRAN-UHFFFAOYSA-N
sodium triethylborohydride, sodium triethylborate, sodium triethylhydroborate, sodium triethylhydridoborate, sodium triethylhydroborate 1-, sodium triethylborohydride solution, sodium triethyl-? 2-boranuide, sodiumtriethylborohydride 1m in toluene
23667700
sodium;triethylboron(1-)
[B-](CC)(CC)CC.[Na+]
17979-81-6, 109-99-9
C6H16BNa
121.99
MFCD00011704
UKYGKELSBAWRAN-UHFFFAOYSA-N
sodium triethylborohydride, sodium triethylborate, sodium triethylhydroborate, sodium triethylhydridoborate, sodium triethylhydroborate 1-, sodium triethylborohydride solution, sodium triethyl-? 2-boranuide, sodiumtriethylborohydride 1m in toluene
23667700
sodium;triethylboron(1-)
[B-](CC)(CC)CC.[Na+]
Reduction reagent
Density 0.8900g/mL
Specific Gravity 0.89
Quantity 800mL
Flash Point −17°C
Packaging AcroSeal™ Glass bottle
Color Colorless
Linear Formula NaB(C2H5)3H
Solubility Information Solubility in water: rzacts.
Formula Weight 121.99
Physical Form Solution
Chemical Name or Material Sodium triethylborohydride, 1M solution in THF
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
In contact with water releases flammable gas.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violentl

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

EINECSNumber : 241-903-1