Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tripolyphosphate, Anhydrous, FCC, 85%, Spectrum™

Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
7758-29-4
Na5O10P3
367.86
HWGNBUXHKFFFIH-UHFFFAOYSA-I
pentasodium bis(phosphonatooxy)phosphinate
[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O
7758-29-4
Na5O10P3
367.86
HWGNBUXHKFFFIH-UHFFFAOYSA-I
pentasodium bis(phosphonatooxy)phosphinate
[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

S1508, 7758-29-4, Na5P3O10

Spectrum™ Sodium Tripolyphosphate, Anhydrous, FCC is a food preservative and as an emulsifier to retain moisture. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 85%
Insoluble Matter 0.001
Lead (Pb) 2 mg/kg
Packaging Poly Pail
Quantity 12 kg