Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium tungstate dihydrate, 99+%, for analysis, ACROS Organics™
Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
Description
Oxidation reagent for analysis
Specifications
Assay Percent Range | 99% min |
Linear Formula | Na2WO4·2H2O |
Fieser | 15,295 |
Merck Index | 15,8826 |
Arsenic (As) | 5ppm max. |
Solubility Information | Solubility in water: 730g/L (20°C). Other solubilities: insoluble in alcohol and acids |
Formula Weight | 329.85 |
Physical Form | Crystalline Powder of Flakes |
Percent Purity | 99+% |
Grade | Analysis |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
GHS Signal Word: Warning
RTECSNumber : YO7900000