Alfa Aesar™ Sodium zirconium oxide, 98%

Manufacturer: Alfa Aesar™ 1876222

Quantity 100g

 View more versions of this product

Catalog No. AA1876222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12201-48-8
    Formula Weight 185.2
    MDL Number MFCD00014245
    Synonym Sodium zirconate
    Chemical Name or Material Sodium zirconium oxide

    Specifications

    Formula Weight 185.2
    TSCA TSCA
    Mesh Size −100 Mesh
    MDL Number MFCD00014245
    Synonym Sodium zirconate
    Color White
    Quantity 100g
    Physical Form Powder
    CAS 12201-48-8
    Chemical Name or Material Sodium zirconium oxide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit