Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ SoftCIDE™ Extra-Mild Antimicrobial Handwash
Effectively eliminate bacteria while replenishing the natural oils in your skin with Epredia™ SoftCIDE™ Extra-Mild Antimicrobial Handwash.
Supplier: Epredia™ 2100412001
This item is not returnable.
View return policy
Description
- Uses 1% PCMX as an active ingredient
- Unique formulation contains emollients
- Effective against a broad spectrum or microorganisms
- Soap is extremely gentle; pH balanced with the skin
Specifications
Extra Mild Anti-microbial Hand Wash | |
Squeeze Bottle | |
118 mL |
Squeeze Bottle | |
4 oz. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction