Learn More
Thermo Scientific™ SoftFit-L™ Filtered Pipette Tips, TipCycle Recycle Kit, Hinged Racks
The pipette tip and rack mail-back program provides a simple and effective solution for recycling PP#5 single use plastics. The TipCycle Kit included with your case of pipette tips to collect used pipette tips and empty racks. Complete waiver, attach pre-paid shipping label and return for recycling.
Supplier: Thermo Scientific™ 2789RCHR
Description
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
- Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
- Complete TipCycle Pipette Tip Recycling Kit Provides: 1 case of pipette tips, instructions for return, a box liner and liner tie help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, FedEx return shipping label adhered to top of recycle package, then simply ship the recycle package.
- Hinged rack provides easy one-hand access to tips
- Color-coded inserts allow quick identification of tip volume
- Sterile tips certified free: DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin
Recommended for:
- Liquid Handling
- Molecular Biology
- PCR
- Biotechnology
- Front latch closes and locks, ensuring safe and secure storage throughout
- Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
- Secure snap design securely holds insert in place, preventing it from being lifted out while pipetting
- Lid rim prevents racks from sliding around when stacked and carted around a lab
Specifications
1200 μL | |
Sterile | |
Not Autoclavable | |
3.42 in. | |
DNase, RNase, DNA, ATP, and Endotoxin Free | |
Rainin™ LTS™ LiteTouch™ | |
Specialty | |
Softfit-L |
Filtered | |
Natural | |
1200 μL, Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit | |
8.67 cm | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
Hinged Rack | |
TipCycle, Pipette Tip and Rack Recycling Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.