Thermo Scientific™ SoftGUARD™ Extra-Strength Barrier Hand Cream Available on GSA/VA Contract for Federal Government customers only.
Prevent skin irritation caused by frequent washing and gloving with Thermo Scientific™ SoftGUARD Extra-Strength Barrier Hand Cream.
Manufacturer: Thermo Scientific™ 1102712001
 View more versions of this product
Catalog No. 14-379-21
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Size (English) 27 oz.
    Size (Metric) 798mL
    Packaging Bag-in-a-box carton refill
    Prevent skin irritation caused by frequent washing and gloving. Thermo Scientific™ SoftGUARD Extra-Strength Barrier Hand Cream protects and moisturizes, allowing damaged skin to heal.
    Unique barrier feature helps prevent the occlusion of the skin.
    • Compatible with latex, nitrile, vinyl, PCMX, CHG, and antimicrobial soap—petroleum- and mineral oil-free
    • Utilizes an extra-strength antimicrobial preservative to prevent cross-contamination
    • Absorbs immediately
    • Nongreasy, fragrance-free and pH balanced with the skin
  • Description & Specifications
Related Products
Product Content Feedback