Learn More
Epredia™ SoftGUARD™ Extra-Strength Barrier Hand Cream
Prevent skin irritation caused by frequent washing and gloving with Epredia™ SoftGUARD Extra-Strength Barrier Hand Cream.
Supplier: Epredia™ 1112804001
This item is not returnable.
View return policy
Description
Unique barrier feature helps prevent the occlusion of the skin.- Compatible with latex, nitrile, vinyl, PCMX, CHG, and antimicrobial soap—petroleum- and mineral oil-free
- Utilizes an extra-strength antimicrobial preservative to prevent cross-contamination
- Absorbs immediately
- Nongreasy, fragrance-free and pH balanced with the skin
Specifications
Extra-strength Barrier Hand Cream | |
Refill Jug | |
1 gal. | |
3.8 L |