Learn More
Aviva Systems Biology Corp SOHLH1 Antibody - C-terminal region (ARP54547_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54547_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOHLH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SOHLH1. Alternate names: C9orf157, NOHLH, TEB2, bA100C15.3, bHLHe80ApplicationWB. Species reactivity: Horse, Human, Yeast
Catalog No. 50-014-61241