Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOHLH1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | SOHLH1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18028420
|
Novus Biologicals
NBP18028420UL |
20 μL |
Each for $152.22
|
|
NBP180284
|
Novus Biologicals
NBP180284 |
100 μL |
Each for $436.00
|
|
Description
SOHLH1 Polyclonal specifically detects SOHLH1 in Mouse samples. It is validated for Western Blot.Specifications
SOHLH1 | |
Polyclonal | |
Rabbit | |
NP_001001714 | |
402381 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
bA100C15.3, bHLHe80, C9orf157, NOHLHchromosome 9 open reading frame 157, spermatogenesis and oogenesis specific basic helix-loop-helix 1, spermatogenesis- and oogenesis-specific basic helix-loop-helix-containingprotein 1, TEB2newborn ovary helix loop helix | |
SOHLH1 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title