Learn More
Quidel Solana SARS-CoV-2 Assay
Isothermal Reverse Transcriptase-Helicase-Dependent Amplification (RT-HDA) assay intended for the qualitative detection of nucleic acid from SARS-CoV-2 in nasopharyngeal & nasal swab specimens from individuals suspected of COVID-19 by their healthcare provider.
Specifications
Quantity 96/Kit
Certifications/Compliance Emergency Use Authorization (EUA)
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
23-044-158
View Documents
Quidel
M313
Nasal and Nasopharyngeal
Pack of 96 for $3,918.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-044-159
View Documents
Quidel
M956
Nasopharyngeal
Pack of 96 for $4,115.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-044-161
View Documents
Quidel
M958
Nasal
Pack of 96 for $4,115.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • RT-HDA technology
  • Frozen Master Mix
  • Small, easy-to-use instrument
  • CLIA moderately complex
  • Storage (Process Buffer 2°C to 8°C storage, Master Mix -70°C or below)
  • Room temperature set up
Specifications
Specifications
96/Kit
Emergency Use Authorization (EUA)