Learn More
Soldering flux, liquid, Thermo Scientific Chemicals
All purpose flux for general soldering
$19.65 - $57.00
Specifications
Chemical Name or Material | Soldering flux |
---|---|
Density | 1.26 g/mL |
MDL Number | MFCD01868519 |
UN Number | UN1840 |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA45581AE
|
Thermo Scientific Chemicals
045581AE |
100 mL |
Each
for $19.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA45581AP
|
Thermo Scientific Chemicals
045581AP |
500 mL |
Each for $57.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Soldering flux | |
MFCD01868519 | |
Liquid |
1.26 g/mL | |
UN1840 |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: ZINC CHLORIDE, SOLUTION
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.